Assay Details
| Gene Symbol: COX19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256807.1 | NP_001243736.1 | 2 - 3 | 238 | 119 | |
| GenBank mRNA | BU119719.1 | - | 2 - 3 | 249 | 119 | |
| BU382349.1 | - | 2 - 3 | 211 | 119 | ||
More Information
Set Membership: |
|

COX19
Genomic Map