Assay Details
| Gene Symbol: MFAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256948.1 | NP_001243877.1 | 6 - 7 | 955 | 60 | |
| GenBank mRNA | DR412038.1 | - | 510 | 60 | ||
| M76679.1 | - | 6 - 7 | 940 | 60 | ||
More Information
Set Membership: |
|

MFAP1
Genomic Map