Assay Details
| Gene Symbol: PGRMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271939.1 | NP_001258868.1 | 1 - 2 | 356 | 63 | |
| GenBank mRNA | AJ719868.1 | - | 1 - 2 | 340 | 63 | |
| AJ720052.1 | - | 1 - 2 | 355 | 63 | ||
| CN210800.1 | - | 1 - 2 | 356 | 63 | ||
More Information
Set Membership: |
|

PGRMC1
Genomic Map