Assay Details
| Gene Symbol: SNRPF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277362.1 | NP_001264291.1 | 1 - 2 | 81 | 76 | |
| GenBank mRNA | BX950571.2 | - | 1 - 2 | 65 | 76 | |
| CN228901.1 | - | 1 - 2 | 81 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001277362.1 transcripts.
Set Membership: |
|

SNRPF
Genomic Map