Assay Details
| Gene Symbol: ATP6V0E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277387.1 | NP_001264316.1 | 1 - 2 | 212 | 64 | |
| NM_001277388.1 | NP_001264317.1 | 1 - 2 | 212 | 64 | ||
| GenBank mRNA | BU237585.1 | - | 1 - 2 | 124 | 64 | |
| BU285051.1 | - | 1 - 2 | 74 | 64 | ||
| BU326260.1 | - | 1 - 2 | 221 | 64 | ||
| BX930338.2 | - | 1 - 2 | 121 | 64 | ||
| CR385447.1 | - | 1 - 2 | 112 | 64 | ||
More Information
Set Membership: |
|

ATP6V0E2
Genomic Map