Assay Details
| Gene Symbol: COX6A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277509.1 | NP_001264438.1 | 2 - 3 | 270 | 81 | |
| GenBank mRNA | BX277552.4 | - | 2 - 3 | 270 | 81 | |
More Information
Set Membership: |
|

COX6A1
Genomic Map