Assay Details
| Gene Symbol: CACFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277657.1 | NP_001264586.1 | 5 - 6 | 686 | 59 | |
| XM_025156059.1 | XP_025011827.1 | 8 - 9 | 3596 | 59 | ||
| GenBank mRNA | BX933023.1 | - | 3 - 4 | 291 | 59 | |
| CR385838.1 | - | 5 - 6 | 687 | 59 | ||
More Information
Set Membership: |
|

CACFD1
Genomic Map