Assay Details
| Gene Symbol: UQCRHL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277673.1 | NP_001264602.1 | 2 - 3 | 121 | 65 | |
| GenBank mRNA | BU417752.1 | - | 2 - 3 | 122 | 65 | |
| CN223908.1 | - | 2 - 3 | 87 | 65 | ||
| CR385442.1 | - | 2 - 3 | 146 | 65 | ||
More Information
Set Membership: |
|

UQCRHL
Genomic Map