Assay Details
| Gene Symbol: ATP6V1G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277677.1 | NP_001264606.1 | 1 - 2 | 172 | 96 | |
| GenBank mRNA | BX935024.1 | - | 1 - 2 | 92 | 96 | |
| CR352510.1 | - | 1 - 2 | 109 | 96 | ||
| CV892958.1 | - | 1 - 2 | 174 | 96 | ||
More Information
Set Membership: |
|

ATP6V1G1
Genomic Map