Assay Details
Gene Symbol: TMEM207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277695.1 | NP_001264624.1 | 2 - 3 | 200 | 65 | |
XM_015291405.2 | XP_015146891.1 | 2 - 3 | 276 | 65 | ||
GenBank mRNA | BU373661.1 | - | 2 - 3 | 168 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
