Assay Details
| Gene Symbol: TMEM207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277695.1 | NP_001264624.1 | 3 - 4 | 239 | 62 | |
| GenBank mRNA | BU373661.1 | - | 3 - 4 | 207 | 62 | |
More Information
Set Membership: |
|

TMEM207
Genomic Map