Assay Details
Gene Symbol: FABP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277700.1 | NP_001264629.1 | 1 - 2 | 317 | 76 | |
NM_001277701.1 | NP_001264630.1 | 1 - 2 | 317 | 76 | ||
GenBank mRNA | BU242467.1 | - | 1 - 2 | 317 | 76 | |
BU336217.1 | - | 1 - 2 | 129 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001277700.1, NM_001277701.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
