Assay Details
Gene Symbol: FABP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277700.1 | NP_001264629.1 | 3 - 4 | 506 | 105 | |
NM_001277701.1 | NP_001264630.1 | 3 - 4 | 509 | 105 | ||
XM_015293653.2 | XP_015149139.1 | 3 - 4 | 637 | 105 | ||
GenBank mRNA | BU242467.1 | - | 3 - 4 | 509 | 105 | |
BU336217.1 | - | 3 - 4 | 319 | 105 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001277700.1, NM_001277701.1, XM_015293653.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
