Assay Details
Gene Symbol: FABP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277700.1 | NP_001264629.1 | 4 - 5 | 682 | 78 | |
NM_001277701.1 | NP_001264630.1 | 4 - 5 | 685 | 78 | ||
XM_015293653.2 | XP_015149139.1 | 4 - 5 | 813 | 78 | ||
GenBank mRNA | BU336217.1 | - | 4 - 5 | 495 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
