Assay Details
| Gene Symbol: GPX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277854.2 | NP_001264783.2 | 1 - 2 | 226 | 74 | |
| GenBank mRNA | BX273421.3 | - | 1 - 2 | 52 | 74 | |
| BX273422.4 | - | 1 - 2 | 226 | 74 | ||
More Information
Set Membership: |
|

GPX2
Genomic Map