Assay Details
| Gene Symbol: LSM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277862.1 | NP_001264791.1 | 2 - 3 | 154 | 88 | |
| GenBank mRNA | CD735759.1 | - | 2 - 3 | 155 | 88 | |
| CN234114.1 | - | 2 - 3 | 148 | 88 | ||
More Information
Set Membership: |
|

LSM3
Genomic Map