Assay Details
| Gene Symbol: JDP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277887.1 | NP_001264816.1 | 2 - 3 | 307 | 72 | |
| XM_015287389.2 | XP_015142875.1 | 3 - 4 | 482 | 72 | ||
More Information
Set Membership: |
|

JDP2
Genomic Map