Assay Details
| Gene Symbol: PDXK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278084.3 | NP_001265013.1 | 1 - 2 | 200 | 75 | |
| NM_001278085.1 | NP_001265014.1 | 1 - 2 | 200 | 75 | ||
| XM_025149534.1 | XP_025005302.1 | 1 - 2 | 308 | 75 | ||
| GenBank mRNA | BU425734.1 | - | 1 - 2 | 148 | 75 | |
| BX934150.1 | - | 1 - 2 | 119 | 75 | ||
More Information
Set Membership: |
|

PDXK
Genomic Map