Assay Details
| Gene Symbol: AASDHPPT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278088.1 | NP_001265017.1 | 3 - 4 | 509 | 109 | |
| XM_015278138.2 | XP_015133624.1 | 3 - 4 | 547 | 109 | ||
| GenBank mRNA | AM065627.1 | - | 3 - 4 | 509 | 109 | |
| CF254267.1 | - | 2 - 3 | 280 | 109 | ||
More Information
Set Membership: |
|

AASDHPPT
Genomic Map