Assay Details
| Gene Symbol: SLC9A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285935.2 | NP_001272864.1 | 1 - 2 | 266 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC9A2| Gene Symbol: SLC9A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285935.2 | NP_001272864.1 | 1 - 2 | 266 | 78 | |
Set Membership: |
|