Assay Details
Gene Symbol: MORN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001302147.1 | NP_001289076.1 | 4 - 5 | 313 | 77 | |
XM_004935276.2 | XP_004935333.1 | 4 - 5 | 372 | 77 | ||
XM_025148465.1 | XP_025004233.1 | 3 - 4 | 434 | 77 | ||
GenBank mRNA | BU361298.1 | - | 4 - 5 | 313 | 77 | |
CR523028.1 | - | 1 - 2 | 1014 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
