Assay Details
Gene Symbol: SCRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001302178.1 | NP_001289107.1 | 1 - 2 | 260 | 125 | |
XM_015276237.2 | XP_015131723.1 | 2 - 3 | 125 | |||
GenBank mRNA | CN220233.1 | - | 1 - 2 | 272 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
