Assay Details
| Gene Symbol: SLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302187.1 | NP_001289116.1 | 1 - 2 | 86 | 85 | |
| GenBank mRNA | BX930810.1 | - | 1 - 2 | 115 | 85 | |
| BX935880.1 | - | 1 - 2 | 85 | 85 | ||
| BX935884.1 | - | 1 - 2 | 87 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001302187.1 transcripts.
Set Membership: |
|

SLN
Genomic Map