Assay Details
Gene Symbol: TRIM39.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001312883.1 | NP_001299812.1 | 2 - 3 | 915 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
