Assay Details
| Gene Symbol: GDPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318414.1 | NP_001305343.1 | 3 - 4 | 314 | 77 | |
| GenBank mRNA | BX934885.1 | - | 3 - 4 | 329 | 77 | |
| KC894727.1 | - | 3 - 4 | 314 | 77 | ||
More Information
Set Membership: |
|

GDPD1
Genomic Map