Assay Details
| Gene Symbol: MYH1F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319015.1 | NP_001305944.1 | 11 - 12 | 1265 | 105 | |
| XM_001231408.4 | XP_001231409.1 | 12 - 13 | 1349 | 105 | ||
| GenBank mRNA | AB021180.1 | - | 11 - 12 | 1265 | 105 | |
More Information
Set Membership: |
|

MYH1F
Genomic Map