Assay Details
Gene Symbol: C2orf88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001302144.1 | NP_001289073.1 | 2 | 419 | 87 | |
XM_015289067.2 | XP_015144553.1 | 8 | 1446 | 87 | ||
GenBank mRNA | BM487021.1 | - | 2 | 419 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
