Assay Details
| Gene Symbol: CITED4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204718.2 | NP_990049.1 | 2 | 1814 | 80 | |
| GenBank mRNA | AF261079.1 | - | 1 | 1757 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CITED4| Gene Symbol: CITED4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204718.2 | NP_990049.1 | 2 | 1814 | 80 | |
| GenBank mRNA | AF261079.1 | - | 1 | 1757 | 80 | |
Set Membership: |
|