Assay Details
| Gene Symbol: PDP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199910.1 | NP_001186839.1 | 2 | 3224 | 101 | |
| XM_015282931.2 | XP_015138417.1 | 3 | 8154 | 101 | ||
| XM_015282932.2 | XP_015138418.1 | 2 | 3292 | 101 | ||
More Information
Set Membership: |
|

PDP1
Genomic Map