Assay Details
| Gene Symbol: COA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277642.1 | NP_001264571.1 | 4 | 673 | 58 | |
| NM_001277644.1 | NP_001264573.1 | 2 | 323 | 58 | ||
| GenBank mRNA | BU264356.1 | - | 1 | 220 | 58 | |
| BX929699.1 | - | 2 | 438 | 58 | ||
| BX929802.2 | - | 2 | 423 | 58 | ||
| BX929824.2 | - | 1 | 698 | 58 | ||
| BX930119.1 | - | 3 | 377 | 58 | ||
| BX935415.2 | - | 2 | 244 | 58 | ||
More Information
Set Membership: |
|

COA7
Genomic Map