Assay Details
Gene Symbol: GPX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277853.2 | NP_001264782.2 | 2 | 717 | 100 | |
GenBank mRNA | CR353639.1 | - | 2 | 696 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
