Assay Details
Gene Symbol: LOC107050456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025145463.1 | XP_025001231.1 | 3 - 4 | 391 | 67 | |
GenBank mRNA | BX950841.1 | - | 2 - 3 | 313 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
