Assay Details
| Gene Symbol: PAQR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015276524.2 | XP_015132010.1 | 3 - 4 | 610 | 90 | |
| GenBank mRNA | CR338782.1 | - | 1 - 2 | 215 | 90 | |
More Information
Set Membership: |
|

PAQR3
Genomic Map