Assay Details
Gene Symbol: MED18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003642960.4 | XP_003643008.1 | 1 - 2 | 101 | 64 | |
GenBank mRNA | BX929481.1 | - | 1 - 2 | 118 | 64 | |
BX930683.2 | - | 1 - 2 | 76 | 64 | ||
BX932980.2 | - | 1 - 2 | 122 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



