Assay Details
| Gene Symbol: C14H7orf50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_414763.6 | XP_414763.2 | 4 - 5 | 508 | 81 | |
| GenBank mRNA | BX929499.1 | - | 4 - 5 | 543 | 81 | |
| BX930060.1 | - | 1 - 2 | 46 | 81 | ||
| BX932624.1 | - | 1 - 2 | 502 | 81 | ||
More Information
Set Membership: |
|

C14H7orf50
Genomic Map