Assay Details
Gene Symbol: CA15L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_415218.6 | XP_415218.3 | 2 - 3 | 282 | 101 | |
GenBank mRNA | BX929589.1 | - | 2 - 3 | 192 | 101 | |
CR352958.1 | - | 2 - 3 | 217 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
