Assay Details
| Gene Symbol: VSIG10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015275382.2 | XP_015130868.1 | 6 - 7 | 1221 | 77 | |
| XM_415284.6 | XP_415284.5 | 6 - 7 | 1106 | 77 | ||
| GenBank mRNA | BX929786.1 | - | 3 - 4 | 219 | 77 | |
More Information
Set Membership: |
|

VSIG10
Genomic Map