Assay Details
| Gene Symbol: TTC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_426665.6 | XP_426665.3 | 9 - 10 | 1043 | 67 | |
| GenBank mRNA | CR352818.1 | - | 8 - 9 | 886 | 67 | |
| CR386652.1 | - | 7 - 8 | 812 | 67 | ||
More Information
Set Membership: |
|

TTC4
Genomic Map