Assay Details
| Gene Symbol: ANTXRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015288443.2 | XP_015143929.1 | 15 - 16 | 1307 | 71 | |
| GenBank mRNA | CR352919.1 | - | 3 - 4 | 862 | 71 | |
More Information
Set Membership: |
|

ANTXRL
Genomic Map