Assay Details
| Gene Symbol: C3orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025153777.1 | XP_025009545.1 | 4 - 5 | 472 | 76 | |
| GenBank mRNA | CR353238.1 | - | 1 - 2 | 159 | 76 | |
More Information
Set Membership: |
|

C3orf33
Genomic Map