Assay Details
Gene Symbol: TRIQK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025148041.1 | XP_025003809.1 | 4 | 2928 | 91 | |
GenBank mRNA | CR353414.1 | - | 1 | 516 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
