Assay Details
| Gene Symbol: TUBB2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001231209.5 | XP_001231210.4 | 4 | 2779 | 104 | |
| GenBank mRNA | CR385146.1 | - | 1 | 119 | 104 | |
More Information
Set Membership: |
|

TUBB2A
Genomic Map