Assay Details
Gene Symbol: CCNYL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015289509.2 | XP_015144995.1 | 4 - 5 | 698 | 135 | |
GenBank mRNA | CR385362.1 | - | 3 - 4 | 163 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
