Assay Details
| Gene Symbol: FAM196A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004942352.3 | XP_004942409.3 | 4 | 4119 | 110 | |
| GenBank mRNA | CR385652.1 | - | 1 | 722 | 110 | |
More Information
Set Membership: |
|

FAM196A
Genomic Map