Assay Details
Gene Symbol: LOC426626 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015299879.2 | XP_015155365.1 | 6 - 7 | 795 | 145 | |
XM_015299880.2 | XP_015155366.1 | 6 - 7 | 792 | 145 | ||
GenBank mRNA | CR385917.1 | - | 4 - 5 | 336 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
