Assay Details
| Gene Symbol: CLCN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004938401.3 | XP_004938458.2 | 12 | 5447 | 83 | |
| GenBank mRNA | CR386105.1 | - | 1 | 420 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLCN4| Gene Symbol: CLCN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004938401.3 | XP_004938458.2 | 12 | 5447 | 83 | |
| GenBank mRNA | CR386105.1 | - | 1 | 420 | 83 | |
Set Membership: |
|