Assay Details
| Gene Symbol: CD55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015299057.2 | XP_015154543.1 | 284 | 114 | ||
| XM_417981.6 | XP_417981.4 | 568 | 114 | |||
| GenBank mRNA | CR386356.1 | - | 270 | 114 | ||
More Information
Set Membership: |
|

CD55
Genomic Map