Assay Details
| Gene Symbol: SPATA17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_419414.6 | XP_419414.1 | 3 - 4 | 279 | 91 | |
| GenBank mRNA | CR386787.1 | - | 2 - 3 | 449 | 91 | |
More Information
Set Membership: |
|

SPATA17
Genomic Map