Assay Details
| Gene Symbol: VCPKMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004936493.2 | XP_004936550.1 | 5 - 6 | 844 | 61 | |
| XM_421460.5 | XP_421460.3 | 5 - 6 | 812 | 61 | ||
| GenBank mRNA | CR387534.1 | - | 3 - 4 | 640 | 61 | |
| CR391338.1 | - | 3 - 4 | 310 | 61 | ||
More Information
Set Membership: |
|

VCPKMT
Genomic Map