Assay Details
Gene Symbol: CNNM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004942249.3 | XP_004942306.3 | 3 - 4 | 1978 | 69 | |
XM_426532.6 | XP_426532.5 | 3 - 4 | 1978 | 69 | ||
GenBank mRNA | CR387567.1 | - | 575 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
